Assay Details
Target Gene Details
Entrez Gene ID: | 15184 |
Gene Name: | histone deacetylase 5 |
Gene Aliases: |
AI426555, Hdac4, mHDA1, mKIAA0600 |
Location: |
Chr.11:102195747-102230186 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac5 | NM_001077696.1 | 26 | 3555 | NP_001071164.1 |
NM_001284248.1 | 28 | 4067 | NP_001271177.1 | |
NM_001284249.1 | 27 | 3664 | NP_001271178.1 | |
NM_001284250.1 | 26 | 3552 | NP_001271179.1 | |
NM_010412.3 | 26 | 3552 | NP_034542.3 | |
AF006602.2 | 25 | 3265 | AAD09834.2 | |
AF207748.1 | 25 | 3184 | AAF31418.1 | |
AK122325.2 | 21 | 2594 | BAC65607.2 | |
AK139202.1 | 27 | 3639 | ||
AK145972.1 | 26 | 3518 | ||
AK146483.1 | 26 | 3491 | BAE27204.1 | |
AK155335.1 | 26 | 3526 | BAE33201.1 | |
AK156008.1 | 17 | 5056 | ||
AK157159.1 | 17 | 5056 | ||
AK161956.1 | 6 | 684 | BAE36653.1 | |
AK191790.1 | ||||
AK194337.1 | ||||
AK194720.1 | ||||
AK198049.1 | ||||
AK213915.1 | ||||
AK215682.1 | ||||
AK216114.1 | ||||
AK216995.1 | ||||
AK220038.1 | ||||
BC060609.1 | 25 | 3346 | AAH60609.1 |
More Information
Set Membership: |
Intragenic Exonic |