Assay Details
Target Gene Details
Entrez Gene ID: | 15184 |
Gene Name: | histone deacetylase 5 |
Gene Aliases: |
AI426555, Hdac4, mHDA1, mKIAA0600 |
Location: |
Chr.11:102195747-102230186 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac5 | NM_001077696.1 | NP_001071164.1 | ||
NM_001284248.1 | NP_001271177.1 | |||
NM_001284249.1 | NP_001271178.1 | |||
NM_001284250.1 | NP_001271179.1 | |||
NM_010412.3 | NP_034542.3 | |||
AF006602.2 | AAD09834.2 | |||
AF207748.1 | AAF31418.1 | |||
AK122325.2 | BAC65607.2 | |||
AK139202.1 | ||||
AK145972.1 | ||||
AK146483.1 | BAE27204.1 | |||
AK155335.1 | BAE33201.1 | |||
AK156008.1 | ||||
AK157159.1 | ||||
AK197372.1 | ||||
BC060609.1 | AAH60609.1 |
More Information
Set Membership: |
Intragenic Non-exonic |