Assay Details
Target Gene Details
Entrez Gene ID: | 217214 |
Gene Name: | N-acetylglutamate synthase |
Gene Aliases: |
1700120E20Rik, argA |
Location: |
Chr.11:102145513-102149528 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nags | NM_145829.2 | 3 | 864 | NP_665828.1 |
AF462069.1 | 3 | 820 | AAL86770.1 | |
AJ489814.1 | 3 | 790 | CAD34015.1 | |
AK075765.1 | 2 | 510 | BAC35941.1 | |
BC057990.1 | 3 | 828 | AAH57990.1 |