Assay Details
Target Gene Details
Entrez Gene ID: | 217127 |
Gene Name: | K(lysine) acetyltransferase 7 |
Gene Aliases: |
Hbo1, Hboa, Myst2 |
Location: |
Chr.11:95271853-95310246 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kat7 | NM_001195003.1 | NP_001181932.1 | ||
NM_001195004.1 | NP_001181933.1 | |||
NM_177619.3 | NP_808287.1 | |||
AK147968.1 | BAE28257.1 | |||
AK169458.1 | ||||
AK170389.1 | BAE41763.1 | |||
AK193799.1 | ||||
AK197830.1 | ||||
AK203619.1 | ||||
AK205260.1 | ||||
AK211586.1 | ||||
BC048904.1 | AAH48904.1 | |||
BC057102.1 | AAH57102.1 | |||
DQ076247.1 |
More Information
Set Membership: |
Intragenic Non-exonic |