Assay Details
Target Gene Details
Entrez Gene ID: | 103534 |
Gene Name: | mannoside acetylglucosaminyltransferase 4, isoenzyme B |
Gene Aliases: |
AA407995, GnTIVb |
Location: |
Chr.11:50225335-50235103 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mgat4b | NM_145926.3 | 2 | 500 | NP_666038.3 |
AB053218.1 | 2 | 202 | BAC55019.1 | |
AK155336.1 | 2 | 501 | BAE33202.1 | |
BY729920.1 | 2 | 448 |
More Information
Set Membership: |
Intragenic Exonic |