Assay Details
Target Gene Details
Entrez Gene ID: | 16199 |
Gene Name: | interleukin 9 receptor |
Gene Aliases: |
- |
Location: |
Chr.11:32187544-32205340 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Il9r | NM_001134458.1 | NP_001127930.1 | ||
NM_008374.2 | NP_032400.1 | |||
AK088694.1 | BAC40509.1 | |||
BC111909.1 | AAI11910.1 | |||
M84746.1 | AAA37871.1 |
More Information
Set Membership: |
Intragenic Non-exonic |