Assay Details
Target Gene Details
Entrez Gene ID: | 24083 |
Gene Name: | N-acetyltransferase domain containing 1 |
Gene Aliases: |
AI256713, F3-2, Gm16515, Gtlf3b |
Location: |
Chr.11:60902246-60913792 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Natd1 | NM_025294.5 | NP_079570.1 | ||
AK004720.2 | BAB23503.1 | |||
AK080453.1 | BAC37922.1 | |||
AK089179.1 | BAC40780.1 | |||
BC034332.1 | AAH34332.1 | |||
BC038816.1 | AAH38816.1 | |||
BY196792.1 |