Assay Details
Target Gene Details
Entrez Gene ID: | 27041 |
Gene Name: | GTPase activating protein (SH3 domain) binding protein 1 |
Gene Aliases: |
AI849976, B430204O07, C87777, G3bp, mKIAA4115 |
Location: |
Chr.11:55469752-55500888 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
G3bp1 | NM_013716.2 | NP_038744.1 | ||
AB001927.1 | BAA19469.1 | |||
AK137344.1 | ||||
AK141689.1 | BAE24800.1 | |||
AK164556.1 | 2 | 170 | ||
AK179797.1 | ||||
AK220230.1 | BAD90155.1 | |||
BC021156.1 | AAH21156.1 |
More Information
Set Membership: |
Intragenic Non-exonic |