Assay Details
Target Gene Details
Entrez Gene ID: | 13808 |
Gene Name: | enolase 3, beta muscle |
Gene Aliases: |
Eno-3, MSE |
Location: |
Chr.11:70654763-70662513 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eno3 | NM_001136062.2 | 9 | 1100 | NP_001129534.1 |
NM_001276285.1 | 9 | 1112 | NP_001263214.1 | |
NM_007933.3 | 9 | 1212 | NP_031959.1 | |
AI841640.1 | ||||
AK002485.1 | 9 | 1072 | BAB22137.1 | |
BC013460.1 | 8 | 1084 | AAH13460.1 | |
CT010193.1 | 8 | 1006 | CAJ18401.1 | |
M20745.1 | 6 | 832 | AAA37554.1 | |
X57747.1 | 9 | 1063 | CAA40913.1 | |
X62667.1 | 8 | 1056 | CAA44540.1 |
More Information
Set Membership: |
Intragenic Exonic |