Assay Details
Target Gene Details
Entrez Gene ID: | 66102 |
Gene Name: | chemokine (C-X-C motif) ligand 16 |
Gene Aliases: |
0910001K24Rik, AV290116, BB024863, CXCL16v1, CXCL16v2, SR-PSOX, Zmynd15, b2b498Clo |
Location: |
Chr.11:70454234-70459984 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cxcl16 | NM_023158.6 | NP_075647.3 | ||
AF277001.1 | AAG31754.1 | |||
AF301017.1 | AAG34366.1 | |||
AK003085.1 | ||||
AK009599.1 | BAB26384.1 | |||
AK028875.1 | BAC26166.1 | |||
AK030515.1 | BAC27000.1 | |||
AK150224.1 | BAE29391.1 | |||
AK151330.1 | BAE30309.1 | |||
AK152102.1 | BAE30949.1 | |||
AK152547.1 | BAE31302.1 | |||
AK153192.1 | BAE31792.1 | |||
AK170428.1 | 2 | 764 | ||
BC019961.1 | AAH19961.1 | |||
GQ380016.1 | ||||
GQ380017.1 |
More Information
Set Membership: |
Intragenic Non-exonic |