Assay Details
Target Gene Details
Entrez Gene ID: | 14710 |
Gene Name: | guanine nucleotide binding protein (G protein), gamma transducing activity polypeptide 2 |
Gene Aliases: |
AV096488 |
Location: |
Chr.11:95842219-95845734 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gngt2 | NM_001038664.2 | 3 | 277 | NP_001033753.2 |
NM_001284393.1 | 3 | 281 | NP_001271322.1 | |
NM_001284397.1 | 3 | 278 | NP_001271326.1 | |
NM_023121.2 | 3 | 262 | NP_075610.1 | |
AA028742.1 | 3 | 232 | ||
AA423571.1 | 3 | 151 | ||
AI154009.1 | 3 | 278 | ||
AI159118.1 | 3 | 219 | ||
AK010554.1 | 2 | 1291 | BAB27023.1 | |
BC061005.1 | 3 | 227 | AAH61005.1 | |
BE847751.1 | 3 | 267 | ||
BE994683.1 | 2 | 310 | ||
BF470980.1 | 2 | 91 | ||
BY117112.1 | 3 | 262 | ||
U38500.1 | AAB01731.1 | |||
W35863.1 |
More Information
Set Membership: |
Intragenic Exonic |