Assay Details
Target Gene Details
Entrez Gene ID: | 207165 |
Gene Name: | bromodomain PHD finger transcription factor |
Gene Aliases: |
9430093H17Rik, Falz |
Location: |
Chr.11:107033081-107131922 on Build GRCm38 |
Assay Gene Location: | Within Exon 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bptf | NM_176850.2 | 27 | 8643 | NP_789820.2 |
AK082852.1 | 3 | 213 | BAC38653.1 | |
AK136949.1 | 5 | 902 | ||
AK165902.1 | 7 | 1889 | BAE38448.1 | |
BC021489.1 | 7 | 1816 | AAH21489.1 | |
BC060715.1 | 17 | 3355 | AAH60715.1 | |
GU189417.1 | 27 | 7797 |