Assay Details
Target Gene Details
Entrez Gene ID: | 76184 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 6 |
Gene Aliases: |
6330565N06Rik |
Location: |
Chr.11:110174638-110251776 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abca6 | NM_147218.2 | 24 | 3429 | NP_671751.2 |
AF498361.1 | 24 | 3381 | AAM90907.1 | |
BC132417.1 | 24 | 3266 | ||
BC132419.1 | 24 | 3266 |
More Information
Set Membership: |
Intragenic Exonic |