Assay Details
Target Gene Details
Entrez Gene ID: | 76408 |
Gene Name: | ATP-binding cassette, sub-family C (CFTR/MRP), member 3 |
Gene Aliases: |
1700019L09Rik, ABC31, MLP2, MOAT-D, MRP3 |
Location: |
Chr.11:94343295-94393022 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abcc3 | NM_029600.3 | 21 | 2833 | NP_083876.3 |
AF534127.1 | 21 | 2836 | AAQ10530.1 | |
AF534128.1 | 21 | 2761 | AAQ10531.1 | |
AF534129.1 | 21 | 2836 | AAQ10532.1 | |
AK149551.1 | 21 | 2833 | BAE28953.1 | |
AK155664.1 | 21 | 2811 | BAE33375.1 | |
AK156615.1 | 21 | 3063 | ||
AK172420.1 | 21 | 3063 | ||
AY795569.1 | 21 | 2833 | ||
AY841885.1 | 21 | 2814 | AAX39010.1 | |
BC048825.1 | 21 | 2765 | AAH48825.1 | |
BC150788.1 | 21 | 2827 |
More Information
Set Membership: |
Intragenic Exonic |