Assay Details
Target Gene Details
Entrez Gene ID: | 216227 |
Gene Name: | solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8 |
Gene Aliases: |
BC042593, Vglut3 |
Location: |
Chr.10:89574020-89621249 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc17a8 | NM_182959.3 | NP_892004.1 | ||
AF510321.1 | AAN74643.1 | |||
AK082743.1 | ||||
AK149687.1 | BAE29026.1 | |||
BC042593.1 | AAH42593.1 |
More Information
Set Membership: |
Intragenic Non-exonic |