Assay Details
Target Gene Details
Entrez Gene ID: | 20503 |
Gene Name: | solute carrier family 16 (monocarboxylic acid transporters), member 7 |
Gene Aliases: |
4921534N07Rik, Mct2 |
Location: |
Chr.10:125219284-125389562 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc16a7 | NM_011391.1 | 2 | 276 | NP_035521.1 |
AF058054.1 | 2 | 276 | AAC13719.1 | |
AK076612.1 | 2 | 179 | BAC36415.1 | |
AK085398.1 | 2 | 706 | ||
BC117803.1 | 2 | 121 | ||
BC117804.1 | 2 | 121 | ||
BC126960.1 | 2 | 121 |
More Information
Set Membership: |
Intragenic Exonic |