Assay Details
Target Gene Details
Entrez Gene ID: | 72472 |
Gene Name: | solute carrier family 16 (monocarboxylic acid transporters), member 10 |
Gene Aliases: |
2610103N14Rik, 9830169E08, Mct10, PRO0813, TAT1 |
Location: |
Chr.10:40033532-40142264 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc16a10 | NM_001114332.1 | NP_001107804.1 | ||
NM_028247.4 | NP_082523.1 | |||
AK036743.1 | ||||
AK050229.1 | BAC34135.1 | |||
AK144135.1 | ||||
AK151290.1 | BAE30275.1 | |||
AK151636.1 | ||||
AK151704.1 | BAE30627.1 | |||
AK157355.1 | BAE34061.1 | |||
AK165240.1 | BAE38097.1 | |||
AK165579.1 | BAE38270.1 | |||
BC052877.1 | AAH52877.1 |
More Information
Set Membership: |
Intragenic Non-exonic |