Assay Details
Target Gene Details
Entrez Gene ID: | 50493 |
Gene Name: | thioredoxin reductase 1 |
Gene Aliases: |
TR, TR1, TrxR1 |
Location: |
Chr.10:82833951-82897724 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Txnrd1 | NM_001042513.1 | 12 | 1385 | NP_001035978.1 |
NM_001042514.1 | 11 | 1473 | NP_001035979.1 | |
NM_001042523.1 | 12 | 1602 | NP_001035988.1 | |
NM_015762.2 | 11 | 1278 | NP_056577.2 | |
AB027565.1 | 11 | 1305 | BAA86985.2 | |
AF333036.1 | 12 | 1602 | AAK01140.1 | |
AK146125.1 | 11 | 1279 | BAE26918.1 | |
AK149625.1 | 12 | 1386 | BAE28994.1 | |
AK151927.1 | 12 | 1385 | ||
AK168356.1 | 12 | 1386 | BAE40292.1 | |
AK181640.1 | 2 | 102 | ||
AK206356.1 | 2 | 102 | ||
BC037643.1 | 11 | 1558 | AAH37643.1 |
More Information
Set Membership: |
Intragenic Exonic |