Assay Details
Target Gene Details
Entrez Gene ID: | 103098 |
Gene Name: | solute carrier family 6 (neurotransmitter transporter), member 15 |
Gene Aliases: |
AA536730, AI326450, AI326451, v7-3 |
Location: |
Chr.10:103367458-103419379 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc6a15 | NM_001252330.1 | 5 | 1036 | NP_001239259.1 |
NM_175328.3 | 6 | 1133 | NP_780537.1 | |
AK036136.1 | 5 | 1015 | BAC29315.1 | |
AK045793.1 | 3 | 316 | BAC32493.1 | |
AM085111.1 | 4 | 664 | CAJ29896.1 | |
AY149280.1 | 6 | 1098 | AAN75437.1 | |
AY149281.1 | 5 | 1011 | AAN75438.1 | |
BC076593.1 | 5 | 1036 | AAH76593.1 |
More Information
Set Membership: |
Intragenic Exonic |