Assay Details
Target Gene Details
Entrez Gene ID: | 50493 |
Gene Name: | thioredoxin reductase 1 |
Gene Aliases: |
TR, TR1, TrxR1 |
Location: |
Chr.10:82833951-82897724 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Txnrd1 | NM_001042513.1 | NP_001035978.1 | ||
NM_001042514.1 | NP_001035979.1 | |||
NM_001042523.1 | NP_001035988.1 | |||
NM_015762.2 | NP_056577.2 | |||
AB027565.1 | BAA86985.2 | |||
AF333036.1 | AAK01140.1 | |||
AK011902.1 | BAB27905.1 | |||
AK146125.1 | BAE26918.1 | |||
AK149625.1 | BAE28994.1 | |||
AK151927.1 | ||||
AK168356.1 | BAE40292.1 | |||
AK172383.1 | ||||
AK177272.1 | ||||
AK180432.1 | ||||
AK184063.1 | ||||
AK185936.1 | ||||
AK186760.1 | ||||
AK186863.1 | ||||
AK186897.1 | ||||
AK194169.1 | ||||
AK210892.1 | ||||
BC037643.1 | AAH37643.1 | |||
CJ136679.1 | ||||
CX562215.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |