Assay Details
Target Gene Details
Entrez Gene ID: | 64454 |
Gene Name: | solute carrier family 5 (neutral amino acid transporters, system A), member 4b |
Gene Aliases: |
2010104G07Rik, SAAT1, SGLT3-b, SGLT3b, pSGLT2 |
Location: |
Chr.10:76058618-76111018 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc5a4b | NM_023219.2 | NP_075708.2 | ||
AF251268.1 | AAG01742.1 | |||
AF411960.2 | AAL06342.2 | |||
BC109133.1 | AAI09134.1 | |||
BC109134.1 | AAI09135.1 | |||
BC127612.1 | ||||
BC127615.1 | ||||
BC128465.1 |
More Information
Set Membership: |
Intragenic Non-exonic |