Assay Details
Target Gene Details
Entrez Gene ID: | 320183 |
Gene Name: | methionine sulfoxide reductase B3 |
Gene Aliases: |
D430026P16Rik, I-8-23, MsrB2 |
Location: |
Chr.10:120781098-120899304 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Msrb3 | NM_177092.4 | 6 | 525 | NP_796066.1 |
AK052456.1 | 3 | 138 | ||
AK086975.1 | 6 | 492 | BAC39776.1 | |
AK137163.1 | 6 | 492 | BAE23258.1 | |
BB617432.1 | 6 | 560 | ||
BC096447.1 | 6 | 406 | AAH96447.1 |
More Information
Set Membership: |
Intragenic Exonic |