Assay Details
Target Gene Details
Entrez Gene ID: | 12822 |
Gene Name: | collagen, type XVIII, alpha 1 |
Gene Aliases: |
- |
Location: |
Chr.10:77052178-77166530 on Build GRCm38 |
Assay Gene Location: | Within Exon 40 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Col18a1 | NM_001109991.1 | 38 | 4045 | NP_001103461.1 |
NM_009929.3 | 39 | 3461 | NP_034059.2 | |
AF257775.1 | AAF69009.1 | |||
BC064817.1 | 39 | 3432 | AAH64817.1 | |
BC066080.1 | 39 | 3427 | AAH66080.1 | |
D17546.1 | 37 | 2855 | BAA04483.1 | |
L22545.1 | 37 | 3299 | AAA19787.1 | |
U03714.1 | 9 | 881 | AAA20657.1 |
More Information
Set Membership: |
Intragenic Exonic |