Assay Details
Target Gene Details
Entrez Gene ID: | 12822 |
Gene Name: | collagen, type XVIII, alpha 1 |
Gene Aliases: |
- |
Location: |
Chr.10:77052178-77166530 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Col18a1 | NM_001109991.1 | NP_001103461.1 | ||
NM_009929.3 | NP_034059.2 | |||
AK031798.1 | BAC27554.1 | |||
BC064817.1 | AAH64817.1 | |||
BC066080.1 | AAH66080.1 | |||
D17546.1 | BAA04483.1 | |||
L16898.1 | AAA37434.1 | |||
L22545.1 | AAA19787.1 |
More Information
Set Membership: |
Intragenic Non-exonic |