Assay Details
Target Gene Details
Entrez Gene ID: | 26558 |
Gene Name: | homer homolog 3 (Drosophila) |
Gene Aliases: |
AW146114 |
Location: |
Chr.8:70282891-70294361 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Homer3 | NM_001146153.1 | NP_001139625.1 | ||
NM_011984.2 | NP_036114.2 | |||
AF093261.1 | AAC71025.1 | |||
BC005773.1 | AAH05773.1 | |||
BU054773.1 | ||||
BU707796.1 | ||||
CN539202.1 |