Assay Details
Target Gene Details
Entrez Gene ID: | 67184 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13 |
Gene Aliases: |
2700054G14Rik, AU022060, CDA016, CGI-39, GRIM-19, Grim19 |
Location: |
Chr.8:69887788-69902558 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufa13 | NM_023312.2 | NP_075801.1 | ||
AF286698.1 | AAG28168.1 | |||
AK002999.1 | BAB22505.1 | |||
AK003012.1 | ||||
AK010517.1 | BAB26999.1 | |||
AK012422.1 | BAB28227.1 | |||
BC034899.1 | AAH34899.1 | |||
BC037149.1 | AAH37149.1 | |||
BC055435.1 | AAH55435.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |