Assay Details
Target Gene Details
Entrez Gene ID: | 244349 |
Gene Name: | K(lysine) acetyltransferase 6A |
Gene Aliases: |
1500036M03, 9930021N24Rik, MOZ, Myst3, Zfp220 |
Location: |
Chr.8:22859516-22943262 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kat6a | NM_001081149.1 | 4 | 1145 | NP_001074618.1 |
AK028058.1 | 3 | 1064 | BAC25728.1 | |
AK036885.1 | 4 | 1140 | BAC29621.1 | |
AK039615.1 | 4 | 1143 | BAC30401.1 | |
AK054322.1 | 4 | 1139 | BAC35729.1 | |
AK082463.1 | 4 | 1148 | ||
AK132474.1 | 3 | 1053 | BAE21185.1 | |
AK143408.1 | 4 | 1136 | BAE25366.1 |
More Information
Set Membership: |
Intragenic Exonic |