Assay Details
Target Gene Details
Entrez Gene ID: | 71903 |
Gene Name: | carboxylesterase 2F |
Gene Aliases: |
2310038E17Rik, A430088E12 |
Location: |
Chr.8:104947355-104961130 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ces2f | NM_001079865.2 | NP_001073334.2 | ||
AK040349.1 | ||||
BC117742.1 | ||||
BC117743.1 |
More Information
Set Membership: |
Intragenic Non-exonic |