Assay Details
Target Gene Details
Entrez Gene ID: | 20299 |
Gene Name: | chemokine (C-C motif) ligand 22 |
Gene Aliases: |
ABCD-1, DCBCK, MDC, Scya22 |
Location: |
Chr.8:94745684-94751388 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl22 | NM_009137.2 | 2 | 160 | NP_033163.1 |
AF052505.1 | 2 | 252 | AAC40200.1 | |
AF076596.1 | 2 | 141 | AAD50547.1 | |
AF163476.1 | 2 | 130 | AAD55763.1 | |
AJ238238.1 | 2 | 135 | CAB40894.1 | |
AK155083.1 | 2 | 161 | BAE33035.1 | |
AK155764.1 | 2 | 158 | BAE33426.1 | |
AK156627.1 | 2 | 161 | BAE33782.1 | |
AK170080.1 | 2 | 161 | BAE41551.1 | |
BC012658.1 | 2 | 170 | AAH12658.1 | |
BY198556.1 | 2 | 161 |
More Information
Set Membership: |
Intragenic Exonic |