Assay Details
Target Gene Details
Entrez Gene ID: | 20295 |
Gene Name: | chemokine (C-C motif) ligand 17 |
Gene Aliases: |
Abcd-2, Scya17, Scya17l, Tarc |
Location: |
Chr.8:94807404-94812038 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl17 | NM_011332.3 | NP_035462.2 | ||
AF125570.1 | AAD56600.1 | |||
AF125571.1 | AAD56601.1 | |||
AF125572.1 | AAD56602.1 | |||
AJ242587.1 | CAB45256.1 | |||
AK007663.1 | BAB25171.1 | |||
BC028505.1 | AAH28505.1 |
More Information
Set Membership: |
Intragenic Non-exonic |