Assay Details
Target Gene Details
Entrez Gene ID: | 72254 |
Gene Name: | RIKEN cDNA 1700030K09 gene |
Gene Aliases: |
- |
Location: |
Chr.8:72443880-72460541 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
1700030K09Rik | NM_028170.2 | NP_082446.2 | ||
AK089918.1 | ||||
AK133815.1 | BAE21858.1 | |||
AK139332.1 | ||||
BC008558.1 | AAH08558.1 |