Assay Details
Target Gene Details
Entrez Gene ID: | 109006 |
Gene Name: | cytokine induced apoptosis inhibitor 1 |
Gene Aliases: |
2810413N20Rik, AA617265, AU021794, anamorsin |
Location: |
Chr.8:94819818-94838340 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ciapin1 | NM_134141.4 | 2 | 177 | NP_598902.1 |
AK076116.1 | 2 | 133 | BAC36196.1 | |
AK079615.1 | 2 | 139 | BAC37702.1 | |
AK088757.1 | 1 | 104 | BAC40550.1 | |
AK136821.1 | 2 | 170 | BAE23137.1 | |
AK146282.1 | 2 | 125 | BAE27040.1 | |
AK163705.1 | 2 | 156 | BAE37466.1 | |
AK168252.1 | 2 | 127 | BAE40202.1 | |
AK181039.1 | 1 | 67 | ||
AK186862.1 | 2 | 177 | ||
AK189220.1 | 1 | 67 | ||
AK200007.1 | 1 | 67 | ||
AK220040.1 | 1 | 67 | ||
AY523555.1 | 1 | 98 | AAS09959.1 | |
BC016261.1 | 1 | 95 | AAH16261.1 | |
BC021864.1 | 2 | 117 | AAH21864.1 | |
BY283649.1 | 2 | 196 |