Assay Details
Target Gene Details
Entrez Gene ID: | 234852 |
Gene Name: | charged multivesicular body protein 1A |
Gene Aliases: |
2900018H07Rik, Pcoln3 |
Location: |
Chr.8:123204261-123212788 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Chmp1a | NM_145606.3 | 4 | 304 | NP_663581.1 |
AK046416.1 | 4 | 277 | BAC32719.1 | |
AK159830.1 | 4 | 277 | BAE35409.1 | |
BC010524.1 | 4 | 254 | AAH10524.1 | |
BC023807.1 | 4 | 267 | AAH23807.1 | |
BC036138.1 | 4 | 256 | AAH36138.1 | |
BC036152.1 | 4 | 258 | AAH36152.1 | |
BY112937.1 |
More Information
Set Membership: |
Intragenic Exonic |