Assay Details
Target Gene Details
Entrez Gene ID: | 102466218 |
Gene Name: | microRNA 7054 |
Gene Aliases: |
Gm27557, mmu-mir-7054 |
Location: |
Chr.7:45012426-45012487 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mir7054 | NR_106021.1 |
Target Gene Details
Entrez Gene ID: | 233208 |
Gene Name: | SR-related CTD-associated factor 1 |
Gene Aliases: |
AI480556, SR-A1 |
Location: |
Chr.7:45002948-45016357 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Scaf1 | NM_001008422.1 | NP_001008422.1 | ||
AK141920.1 | BAE24887.1 | |||
BC042576.1 | AAH42576.1 | |||
BC043138.1 | ||||
BC085153.1 | AAH85153.1 |
More Information
Set Membership: |
Intragenic Non-exonic |