Assay Details
Target Gene Details
Entrez Gene ID: | 77582 |
Gene Name: | membrane bound O-acyltransferase domain containing 7 |
Gene Aliases: |
5730589L02Rik, BB1, Leng4, Lpiat, Lpiat1, mBB1 |
Location: |
Chr.7:3677789-3693525 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mboat7 | NM_029934.3 | 7 | 1731 | NP_084210.2 |
AB055410.1 | 7 | 1729 | BAC53808.1 | |
AK019981.1 | 7 | 1142 | BAB31950.1 | |
AK038998.1 | 6 | 2841 | ||
AK150010.1 | 7 | 1125 | BAE29235.1 | |
AK154606.1 | 7 | 1730 | BAE32708.1 | |
AK158998.1 | ||||
AK170124.1 | 7 | 1208 | BAE41580.1 | |
BC023417.1 | 7 | 1179 | AAH23417.1 | |
BC118958.2 | 7 | 1376 | ||
EU016380.1 | 6 | 943 |
More Information
Set Membership: |
Intragenic Exonic |