Assay Details
Target Gene Details
Entrez Gene ID: | 211548 |
Gene Name: | nodal modulator 1 |
Gene Aliases: |
D7Ertd156e, Nomo, pM5 |
Location: |
Chr.7:46033696-46084212 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nomo1 | NM_153057.4 | 14 | 1712 | NP_694697.3 |
AK030584.1 | 14 | 1679 | ||
AK035957.1 | 14 | 1683 | ||
AK080754.1 | 14 | 1575 | ||
AK082495.1 | 14 | 1591 | BAC38508.1 | |
AK082963.1 | 14 | 1571 | BAC38713.1 | |
AK142338.1 | 14 | 1682 | ||
AK150107.1 | 14 | 1666 | ||
AK165643.1 | 14 | 1575 | ||
BC033923.1 | 3 | 296 | AAH33923.1 | |
BC072630.1 | 14 | 1658 | AAH72630.1 |