Assay Details
Target Gene Details
Entrez Gene ID: | 68695 |
Gene Name: | HD domain containing 3 |
Gene Aliases: |
1110033O09Rik, C86475, MESH1 |
Location: |
Chr.7:80343083-80346097 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hddc3 | NM_026812.2 | 3 | 207 | NP_081088.1 |
AK004078.1 | 3 | 208 | BAB23158.1 | |
BC038310.1 | 3 | 220 | AAH38310.1 |
More Information
Set Membership: |
Intragenic Exonic |