Assay Details
Target Gene Details
Entrez Gene ID: | 56347 |
Gene Name: | eukaryotic translation initiation factor 3, subunit C |
Gene Aliases: |
110kDa, 3230401O13Rik, Eif3s8, NIPIL(A3), NipilA3, Xs, Xsl |
Location: |
Chr.7:126546911-126566366 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif3c | NM_146200.1 | 15 | 1684 | NP_666312.1 |
AK088076.1 | 15 | 1727 | ||
AK152693.1 | 15 | 1702 | ||
AK160078.1 | 15 | 1704 | BAE35610.1 | |
AK168160.1 | 15 | 1688 | ||
AK168174.1 | 15 | 1688 | ||
BC024855.1 | 15 | 1684 | AAH24855.1 | |
BC025032.1 | 15 | 1705 | AAH25032.1 | |
BC080763.1 | 2 | 99 | AAH80763.1 | |
U67328.1 | 4 | 456 | AAB39607.1 |
More Information
Set Membership: |
Intragenic Exonic |