Assay Details
Target Gene Details
Entrez Gene ID: | 11772 |
Gene Name: | adaptor-related protein complex 2, alpha 2 subunit |
Gene Aliases: |
2410074K14Rik, AF006990, AW146353, Adtab, C78001, L25 |
Location: |
Chr.7:141562180-141633011 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ap2a2 | NM_007459.3 | NP_031485.3 | ||
AK088500.1 | BAC40392.1 | |||
AK128972.1 | ||||
AK152463.1 | BAE31240.1 | |||
AK173054.1 | BAD32332.1 | |||
BC010597.1 | AAH10597.1 | |||
BC058099.1 | AAH58099.1 | |||
X14972.1 | CAA33097.1 |
Target Gene Details
Entrez Gene ID: | 100036523 |
Gene Name: | predicted gene, 16982 |
Gene Aliases: |
- |
Location: |
Chr.7:141610066-141613747 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gm16982 | NR_040337.1 | 4 | 1314 | |
AK016549.1 | 4 | 1315 | BAB30297.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |