Assay Details
Target Gene Details
Entrez Gene ID: | 20392 |
Gene Name: | sarcoglycan, epsilon |
Gene Aliases: |
e-SG |
Location: |
Chr.6:4674350-4747204 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sgce | NM_001130188.1 | 1 | 110 | NP_001123660.1 |
NM_001130189.1 | 1 | 110 | NP_001123661.1 | |
NM_001130190.1 | 1 | 110 | NP_001123662.1 | |
NM_001130191.1 | 1 | 110 | NP_001123663.1 | |
NM_011360.3 | 1 | 110 | NP_035490.3 | |
AK132210.1 | BAE21034.1 | |||
BC012665.1 | AAH12665.1 | |||
BY336674.1 | 1 | 113 |
More Information
Set Membership: |
Intragenic Exonic |