Assay Details
Target Gene Details
Entrez Gene ID: | 101206 |
Gene Name: | transcriptional adaptor 3 |
Gene Aliases: |
1110004B19Rik, ADA3, AI987856, Tada3l |
Location: |
Chr.6:113366633-113377939 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tada3 | NM_133932.2 | NP_598693.1 | ||
AF383154.1 | AAM21463.1 | |||
AK078906.1 | BAC37451.1 | |||
AK211996.1 | ||||
BC022707.1 | AAH22707.1 | |||
BC026630.1 | AAH26630.1 | |||
BC032195.1 | AAH32195.1 |
More Information
Set Membership: |
Intragenic Non-exonic |