Assay Details
Target Gene Details
Entrez Gene ID: | 330260 |
Gene Name: | paraoxonase 2 |
Gene Aliases: |
6330405I24Rik, AI481612 |
Location: |
Chr.6:5264620-5298373 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pon2 | NM_183308.2 | NP_899131.1 | ||
AK007350.1 | ||||
AK146311.1 | BAE27066.1 | |||
AK152351.1 | ||||
AK171926.1 | BAE42735.1 | |||
BC021887.1 | 5 | 519 | AAH21887.1 | |
BC037140.1 | AAH37140.1 | |||
BC055896.1 | AAH55896.1 | |||
BC062200.1 | AAH62200.1 | |||
L48514.1 | AAC42089.1 |