Assay Details
Target Gene Details
Entrez Gene ID: | 22793 |
Gene Name: | zyxin |
Gene Aliases: |
9530098H06Rik, R75157 |
Location: |
Chr.6:42349621-42360213 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zyx | NM_001289617.1 | 3 | 656 | NP_001276546.1 |
NM_001289618.1 | 3 | 656 | NP_001276547.1 | |
NM_001289619.1 | 3 | 656 | NP_001276548.1 | |
NM_011777.3 | 3 | 656 | NP_035907.1 | |
AK031415.1 | 3 | 454 | BAC27394.1 | |
AK035743.1 | 3 | 427 | BAC29173.1 | |
AK076491.1 | 3 | 427 | ||
AK090110.1 | 3 | 423 | ||
AK147812.1 | 3 | 657 | BAE28154.1 | |
AK161709.1 | 3 | 452 | ||
AK165290.1 | 2 | 590 | ||
AK170571.1 | 3 | 430 | BAE41886.1 | |
AK192670.1 | 1 | 150 | ||
AK205275.1 | 2 | 235 | ||
BC054775.1 | 3 | 615 | AAH54775.1 | |
X99063.1 | 3 | 614 | CAA67510.1 | |
Y07711.1 | 2 | 364 | CAA68984.1 |
More Information
Set Membership: |
Intragenic Exonic |