Assay Details
Target Gene Details
Entrez Gene ID: | 232164 |
Gene Name: | poly(A) binding protein interacting protein 2B |
Gene Aliases: |
- |
Location: |
Chr.6:83806058-83831741 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Paip2b | NM_146169.2 | NP_666281.2 | ||
AK045922.1 | BAC32533.1 | |||
AK159287.1 | BAE34963.1 | |||
AK168804.1 | BAE40634.1 | |||
AK173114.1 | BAD32392.1 | |||
BC017133.1 | AAH17133.2 | |||
BC034096.1 | AAH34096.2 | |||
BC039969.1 | ||||
BC147239.1 | ||||
BC147240.1 | ||||
BY262990.1 |