Assay Details
Target Gene Details
Entrez Gene ID: | 93673 |
Gene Name: | camello-like 2 |
Gene Aliases: |
- |
Location: |
Chr.6:85865422-85869137 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cml2 | NM_053096.3 | NP_444326.2 | ||
AF163315.1 | AAF80484.1 | |||
AK050449.1 | BAC34261.1 | |||
BC039773.1 | AAH39773.1 | |||
JX420287.1 |
More Information
Set Membership: |
Intragenic Non-exonic |