Assay Details
Target Gene Details
Entrez Gene ID: | 213819 |
Gene Name: | CAS1 domain containing 1 |
Gene Aliases: |
Cas1, Cast1 |
Location: |
Chr.6:4600563-4643381 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Casd1 | NM_145398.2 | NP_663373.2 | ||
AB091829.1 | BAC77246.1 | |||
AK193530.1 | ||||
AK200014.1 | ||||
BC018542.1 | AAH18542.1 | |||
BC038009.1 | AAH38009.1 | |||
BC058953.1 | AAH58953.1 | |||
BC125377.1 | ||||
BC125379.1 |
More Information
Set Membership: |
Intragenic Non-exonic |