Assay Details
Target Gene Details
Entrez Gene ID: | 76507 |
Gene Name: | amine oxidase, copper-containing 1 |
Gene Aliases: |
1600012D06Rik, Abp1, DAO |
Location: |
Chr.6:48895255-48909188 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aoc1 | NM_001161621.1 | 1 | 94 | NP_001155093.1 |
NM_001161622.1 | NP_001155094.1 | |||
NM_029638.2 | NP_083914.2 | |||
AK144567.1 | ||||
AK146078.1 | 1 | 95 | BAE26882.1 | |
BB868807.1 | ||||
BC021880.1 | AAH21880.1 | |||
BC034215.1 | AAH34215.1 | |||
BY131896.1 |
More Information
Set Membership: |
Intragenic Exonic |