Assay Details
Target Gene Details
Entrez Gene ID: | 13835 |
Gene Name: | Eph receptor A1 |
Gene Aliases: |
5730453L17Rik, AL033318, Eph, Esk |
Location: |
Chr.6:42358487-42373310 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Epha1 | NM_023580.4 | NP_076069.2 | ||
AF131197.1 | AAG12206.1 | |||
AK028478.1 | BAC25971.1 | |||
BC071215.1 | AAH71215.1 |
Target Gene Details
Entrez Gene ID: | 22793 |
Gene Name: | zyxin |
Gene Aliases: |
9530098H06Rik, R75157 |
Location: |
Chr.6:42349621-42360213 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Zyx | NM_001289617.1 | 11 | 2116 | NP_001276546.1 |
AK031415.1 | 11 | 1914 | BAC27394.1 | |
AK035743.1 | 11 | 1887 | BAC29173.1 |
More Information
Set Membership: |
Intragenic Exonic Non-exonic |