Assay Details
Target Gene Details
Entrez Gene ID: | 66993 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3 |
Gene Aliases: |
1500001J14Rik, 2210409C08Rik, BAF60C |
Location: |
Chr.5:24592622-24624456 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smarcd3 | NM_025891.3 | 11 | 1381 | NP_080167.3 |
AK005094.1 | 11 | 1328 | BAB23813.1 | |
AK019374.1 | 4 | 462 | BAB31685.2 | |
BC013122.1 | 11 | 1418 | AAH13122.2 | |
BC060525.1 | 11 | 1280 | AAH60525.1 |
More Information
Set Membership: |
Intragenic Exonic |