Assay Details
Target Gene Details
Entrez Gene ID: | 78829 |
Gene Name: | TSC22 domain family, member 4 |
Gene Aliases: |
0610009M14Rik, 1700023B23Rik, AI415410, Thg-1pit, Tilz2 |
Location: |
Chr.5:137745969-137768453 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tsc22d4 | NM_023910.6 | NP_076399.4 | ||
NM_029805.1 | NP_084081.1 | |||
AF201286.1 | AAG41219.1 | |||
AK018735.2 | BAB31377.1 | |||
AK040534.1 | BAC30619.1 | |||
AK079510.1 | BAC37664.1 | |||
AK080828.1 | ||||
AK143608.1 | BAE25460.1 | |||
AK144806.1 | ||||
AK150043.1 | ||||
AK154153.1 | BAE32411.1 | |||
AK154490.1 | BAE32622.1 | |||
AK155712.1 | BAE33397.1 | |||
AK170487.1 | BAE41829.1 | |||
BC018544.1 | AAH18544.1 | |||
BC023761.1 | AAH23761.1 | |||
BC145721.1 | ||||
BC145723.1 |